siRNA Details
| Gene Symbol: 1700123O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045185.1 | 2, 3 | 230 | |||
| GenBank mRNA | AK007260.1 | - | 2, 3 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700123O12Rik| Gene Symbol: 1700123O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045185.1 | 2, 3 | 230 | |||
| GenBank mRNA | AK007260.1 | - | 2, 3 | 231 | ||