siRNA Details
| Gene Symbol: Prox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086375.1 | - | 1 | 96 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662944.1 | - | 1 | 96 | Chr. 12 - 85086374 - 85087707 [+] on Build GRCm38 | |

Prox2
Genomic Map