siRNA Details
| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031402.1 | - | 1 | 3687 | ||
| AK079052.1 | - | 1 | 3705 | |||
| AK163653.1 | - | 1 | 2122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcpip1| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031402.1 | - | 1 | 3687 | ||
| AK079052.1 | - | 1 | 3705 | |||
| AK163653.1 | - | 1 | 2122 | |||