siRNA Details
| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144818.1 | - | 1 | 2604 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB817085.1 | - | 1 | 225 | Chr. 4 - 44526386 - 44526781 [-] on Build GRCm38 | |

Pax5
Genomic Map