siRNA Details
| Gene Symbol: 1700062C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006860.1 | - | 2 | 269 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706847.1 | - | 2 | 265 | Chr. 14 - 16000798 - 16037268 [-] on Build GRCm38 | |

1700062C10Rik
Genomic Map