siRNA Details
| Gene Symbol: 4930521E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040602.1 | 1, 2 | 15 | |||
| GenBank mRNA | AK015860.1 | - | 1, 2 | 16 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715411.1 | - | 1, 2 | 13 | Chr. 1 - 44739726 - 44744947 [-] on Build GRCm38 | |

4930521E06Rik
Genomic Map