siRNA Details
| Gene Symbol: 1700113B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007192.1 | - | 1 | 344 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707278.1 | - | 1 | 342 | Chr. 1 - 138261648 - 138262063 [+] on Build GRCm38 | |

1700113B19Rik
Genomic Map