siRNA Details
| Gene Symbol: 4930584F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015867.1 | - | 3 | 541 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715418.1 | - | 3 | 541 | Chr. 5 - 26474935 - 26493289 [-] on Build GRCm38 | |

4930584F24Rik
Genomic Map