siRNA Details
| Gene Symbol: 4930480C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015596.1 | - | 1 | 182 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715146.1 | - | 182 | Chr. - - [-] on Build | ||

4930480C01Rik
Genomic Map