siRNA Details
| Gene Symbol: Gm29805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105054.1 | 1 | 31 | |||
| GenBank mRNA | AK045951.1 | - | 1 | 32 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB644499.1 | - | 1 | 27 | Chr. 16 - 92377498 - 92382679 [-] on Build GRCm38 | |

Gm29805
Genomic Map