siRNA Details
| Gene Symbol: Mettl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012721.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711843.1 | - | 1 | 70 | Chr. 10 - 127045871 - 127046363 [+] on Build GRCm38 | |

Mettl1
Genomic Map