siRNA Details
| Gene Symbol: 4921515E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045711.1 | 8 | 1543 | |||
| GenBank mRNA | AK132982.1 | - | 8 | 1544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921515E04Rik| Gene Symbol: 4921515E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045711.1 | 8 | 1543 | |||
| GenBank mRNA | AK132982.1 | - | 8 | 1544 | ||