siRNA Details
| Gene Symbol: 4921515E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045711.1 | 6 | 1416 | |||
| GenBank mRNA | AK132982.1 | - | 6 | 1417 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921515E04Rik| Gene Symbol: 4921515E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045711.1 | 6 | 1416 | |||
| GenBank mRNA | AK132982.1 | - | 6 | 1417 | ||