siRNA Details
| Gene Symbol: 4921515E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045711.1 | 9 | 1888 | |||
| GenBank mRNA | AK132982.1 | - | 9 | 1889 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV269390.2 | - | 2 | 255 | Chr. 15 - 18078142 - 18165866 [-] on Build GRCm38 | |

4921515E04Rik
Genomic Map