siRNA Details
| Gene Symbol: 4931407E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016440.1 | - | 2 | 316 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716097.1 | - | 2 | 316 | Chr. 14 - 21869266 - 21889766 [-] on Build GRCm38 | |

4931407E12Rik
Genomic Map