siRNA Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018405.1 | - | 1 | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720528.1 | - | 1 | 162 | Chr. 2 - 34785801 - 34786550 [-] on Build GRCm38 |
