siRNA Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018405.1 | - | 1 | 299 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720528.1 | - | 1 | 294 | Chr. 2 - 34785801 - 34786550 [-] on Build GRCm38 | |

Rabepk
Genomic Map