siRNA Details
| Gene Symbol: 8430413D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018407.1 | - | 1 | 769 | ||
| AK048170.1 | - | 1 | 1780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430413D17Rik| Gene Symbol: 8430413D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018407.1 | - | 1 | 769 | ||
| AK048170.1 | - | 1 | 1780 | |||