siRNA Details
| Gene Symbol: 8430413B15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018406.1 | - | 1 | 371 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720529.1 | - | 1 | 371 | Chr. 6 - 73050646 - 73051173 [+] on Build GRCm38 | |

8430413B15Rik
Genomic Map