siRNA Details
| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007997.1 | - | 1 | 365 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707994.1 | - | 1 | 362 | Chr. X - 103493580 - 103494316 [+] on Build GRCm38 | |

Jpx
Genomic Map