siRNA Details
| Gene Symbol: Gm19835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138371.1 | - | 709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19835| Gene Symbol: Gm19835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138371.1 | - | 709 | |||