siRNA Details
| Gene Symbol: Gm32273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085773.1 | - | 1 | 700 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG694997.1 | - | 1 | 133 | Chr. 11 - 68957550 - 68962492 [+] on Build GRCm38 | |

Gm32273
Genomic Map