siRNA Details
| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK172141.1 | - | 1038 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY765891.1 | - | 1 | 530 | Chr. 10 - 56116866 - 56117573 [+] on Build GRCm38 | |

Msl3l2
Genomic Map