siRNA Details
| Gene Symbol: A930005G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044281.1 | - | 1 | 567 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB591645.2 | - | 1 | 565 | Chr. 5 - 115421831 - 115422498 [-] on Build GRCm38 | |

A930005G22Rik
Genomic Map