siRNA Details
| Gene Symbol: Gm28856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087224.1 | - | 2059 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm28856| Gene Symbol: Gm28856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087224.1 | - | 2059 | |||