siRNA Details
Gene Symbol: Gm11538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108029.1 | 2 | 1329 | |||
GenBank mRNA | AK078606.1 | - | 2 | 1330 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108029.1 | 2 | 1329 | |||
GenBank mRNA | AK078606.1 | - | 2 | 1330 |