siRNA Details
| Gene Symbol: Gm16973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046209.1 | 3 | 854 | |||
| NR_046210.1 | 3 | 786 | ||||
| GenBank mRNA | AK045225.1 | - | 3 | 1054 | ||
| BC027359.1 | - | 2 | 660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16973| Gene Symbol: Gm16973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046209.1 | 3 | 854 | |||
| NR_046210.1 | 3 | 786 | ||||
| GenBank mRNA | AK045225.1 | - | 3 | 1054 | ||
| BC027359.1 | - | 2 | 660 | |||