siRNA Details
| Gene Symbol: 2310043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045757.1 | 3 | 593 | |||
| GenBank mRNA | AK009791.1 | - | 3 | 594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310043O21Rik| Gene Symbol: 2310043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045757.1 | 3 | 593 | |||
| GenBank mRNA | AK009791.1 | - | 3 | 594 | ||