siRNA Details
| Gene Symbol: 2310043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045757.1 | 2 | 360 | |||
| GenBank mRNA | AK009791.1 | - | 2 | 361 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709413.1 | - | 2 | 361 | Chr. 15 - 38549391 - 38594934 [-] on Build GRCm38 | |

2310043O21Rik
Genomic Map