siRNA Details
| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021161.1 | - | 1 | 944 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB321505.1 | - | 1 | 260 | Chr. 4 - 118569725 - 118570363 [+] on Build GRCm38 | |

Cfap57
Genomic Map