siRNA Details
| Gene Symbol: 2310074N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010155.1 | - | 1 | 294 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709669.1 | - | 1 | 287 | Chr. 5 - 24447914 - 24448484 [+] on Build GRCm38 | |

2310074N15Rik
Genomic Map