siRNA Details
| Gene Symbol: 1110059G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004311.1 | - | 1 | 654 | ||
| AV235558.1 | - | 1 | 171 | |||
| BB766817.1 | - | 1 | 257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110059G02Rik| Gene Symbol: 1110059G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004311.1 | - | 1 | 654 | ||
| AV235558.1 | - | 1 | 171 | |||
| BB766817.1 | - | 1 | 257 | |||