siRNA Details
| Gene Symbol: 4930467K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045353.1 | 3 | 333 | |||
| GenBank mRNA | AK015522.1 | - | 3 | 333 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715070.1 | - | 3 | 330 | Chr. 10 - 57478381 - 57486342 [-] on Build GRCm38 | |

4930467K11Rik
Genomic Map