siRNA Details
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166127.1 | - | 4 | 641 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734500.1 | - | 4 | 641 | Chr. 5 - 13125414 - 13308959 [+] on Build GRCm38 |
