siRNA Details
| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102283.1 | 5 | 2074 | |||
| NR_102287.1 | 4 | 1098 | ||||
| GenBank mRNA | AK030886.1 | - | 4 | 1918 | ||
| BC064108.1 | - | 2 | 753 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI733270.1 | - | 1 | 85 | Chr. 4 - 155408416 - 155409086 [-] on Build GRCm38 | |

5830444B04Rik
Genomic Map