siRNA Details
| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102283.1 | 5 | 1538 | |||
| NR_102287.1 | 4 | 562 | ||||
| GenBank mRNA | AA119566.1 | - | 3 | 367 | ||
| AK030886.1 | - | 4 | 1382 | |||
| BC064108.1 | - | 2 | 217 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830444B04Rik| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102283.1 | 5 | 1538 | |||
| NR_102287.1 | 4 | 562 | ||||
| GenBank mRNA | AA119566.1 | - | 3 | 367 | ||
| AK030886.1 | - | 4 | 1382 | |||
| BC064108.1 | - | 2 | 217 | |||