siRNA Details
| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102283.1 | 4 | 1250 | |||
| GenBank mRNA | AK030886.1 | - | 3 | 1094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830444B04Rik| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102283.1 | 4 | 1250 | |||
| GenBank mRNA | AK030886.1 | - | 3 | 1094 | ||