siRNA Details
| Gene Symbol: 1700108F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133110.1 | - | 1 | 1836 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV488190.1 | - | 1 | 177 | Chr. 14 - 76679237 - 76679754 [+] on Build GRCm38 | |

1700108F19Rik
Genomic Map