siRNA Details
| Gene Symbol: 4921519G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014924.1 | - | 1 | 486 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714442.1 | - | 1 | 481 | Chr. 10 - 42855709 - 42856679 [-] on Build GRCm38 | |

4921519G19Rik
Genomic Map