siRNA Details
| Gene Symbol: Arhgap32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014385.1 | - | 1 | 719 | ||
| CN669722.1 | - | 1 | 443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap32| Gene Symbol: Arhgap32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014385.1 | - | 1 | 719 | ||
| CN669722.1 | - | 1 | 443 | |||