siRNA Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053064.1 | - | 1 | 2007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst8| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053064.1 | - | 1 | 2007 | ||