siRNA Details
| Gene Symbol: 4930444M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045660.1 | 4 | 1578 | |||
| GenBank mRNA | AK015383.1 | - | 4 | 1579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930444M15Rik| Gene Symbol: 4930444M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045660.1 | 4 | 1578 | |||
| GenBank mRNA | AK015383.1 | - | 4 | 1579 | ||