siRNA Details
| Gene Symbol: Gm19272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029919.1 | - | 3 | 561 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616303.1 | - | 3 | 560 | Chr. 6 - 7301908 - 7305120 [+] on Build GRCm38 | |

Gm19272
Genomic Map