siRNA Details
| Gene Symbol: 1700066D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006901.1 | - | 1 | 689 | ||
| AK018981.1 | - | 1 | 698 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700066D14Rik| Gene Symbol: 1700066D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006901.1 | - | 1 | 689 | ||
| AK018981.1 | - | 1 | 698 | |||