siRNA Details
| Gene Symbol: 1700123J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018982.1 | - | 1 | 82 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707352.1 | - | 1 | 72 | Chr. 12 - 77061026 - 77061247 [-] on Build GRCm38 | |

1700123J03Rik
Genomic Map