siRNA Details
| Gene Symbol: 4832441B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014650.1 | - | 1 | 1197 | ||
| AK029342.1 | - | 1 | 1197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4832441B07Rik| Gene Symbol: 4832441B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014650.1 | - | 1 | 1197 | ||
| AK029342.1 | - | 1 | 1197 | |||