siRNA Details
| Gene Symbol: 4833403J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014653.1 | - | 1 | 879 | ||
| BB168546.1 | - | 1 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833403J16Rik| Gene Symbol: 4833403J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014653.1 | - | 1 | 879 | ||
| BB168546.1 | - | 1 | 194 | |||