siRNA Details
| Gene Symbol: 4833403J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014653.1 | - | 1 | 574 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714150.1 | - | 1 | 574 | Chr. 6 - 48755338 - 48756124 [+] on Build GRCm38 | |

4833403J16Rik
Genomic Map