siRNA Details
| Gene Symbol: 4833406M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014658.1 | - | 1 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833406M21Rik| Gene Symbol: 4833406M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014658.1 | - | 1 | 229 | ||