siRNA Details
| Gene Symbol: 9330133O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045696.1 | 1 | 221 | |||
| GenBank mRNA | AK033985.1 | - | 1 | 162 | ||
| AK042662.1 | - | 1 | 186 | |||
| AK049744.1 | - | 1 | 222 | |||
| AK085519.1 | - | 1 | 172 | |||
| BC094355.1 | - | 1 | 107 | |||

9330133O14Rik
Genomic Map